Learn More
Sklar Instruments Graves Vaginal Speculum, Sklar, Extra Long, 6 in. x 1-1/2 in., Straight, Stainless Steel. EA/1.
Supplier: Sklar Instruments 903803
Graves Vaginal Speculum, Sklar, Extra Long, 6 in. x 1-1/2 in., Straight, Stainless Steel. The Sklar Graves Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix. It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient. This product is an extra long speculum with 6 in. X 1-1/2 in. blades.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.