Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DeltaBiologicals Grass Frog
The grass frog is the ideal dissection specimen for anatomical studies and for introducing students to all the major organ systems.
Supplier: DeltaBiologicals 136.1679.10
This item is not returnable.
View return policy
Description
- Scientific Name = Rana forreri
- Size = 3 to 5 in.
- Pail Pac or Vac-Pac Packaging
- Product is stored and shipped in Delta-Sol, a non-toxic preservative fluid
Specifications
Delta Solution | |
Frog Dissection Specimen | |
10 Pk. | |
3 in. to 5 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction