Learn More
Sklar Instruments Graduated Measure, Sklar, Stainless Steel. It has a 5-1/2 in diameter base and a 6-1/2 in opening. This pitcher will hold up to 64 fluid oz. EA/1.
Supplier: Sklar Instruments 101674
Graduated Measure, Sklar, Stainless Steel. Sklar's Stainless Steel Graduated Measure is a container used to accurately measure fluids or measure and dilute chemicals. The pitcher features cubic centimeter and ounce markings. It has a 5-1/2 inch diameter base and a 6-1/2 inch diameter opening. This pitcher will hold up to 64 fluid ounces.
This item is not returnable.
View return policy