Learn More
Sklar Instruments Graduated Measure, Sklar, Stainless Steel. This product measures 4-3/4 in. x 5-1/4 in. and has a capacity of 32 ounces. EA/1.
SDP
Supplier:  Sklar Instruments 101510
Encompass_Preferred
Graduated Measure, Sklar, Stainless Steel. Sklar's Graduated Measure is a container used to accurately measure fluids or measure and dilute chemicals for use. It resembles a measuring cup and may be referred to as simply a in.measure in. or a in.graduate. in. Calibrations for cubic centimeters and ounces are indicated on the inside of the product. This product measures 4-3/4 in. x 5-1/4 in. and has a capacity of 32 ounces.
Catalog No. 50-143-7022