Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ GR-2 Septa
Designed for routine, isothermal use
Supplier: MilliporeSigma™ Supelco™ 20712
Description
- Septa are very important components of a syringe injection system
- Typically fabricated from elastomeric, self-sealing materials such as silicone rubber, neoprene, or fluoroelastomers
- Suitable to conveniently introducing the sample without causing the system to leak
- Improper septa decompose under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram
- These low cost, gray silicone rubber septa are designed for routine, isothermal use
- Inlet Temperature Range: 50°C to 200°C
Specifications
GR-2 Septum | |
3/16 in. | |
Gray | |
Silicone Rubber |
200°C | |
5 mm | |
Routine, Isothermal Use | |
100 Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction