Learn More
Med Vet International AngelStat Surgical Gown, Blockade, 1-Ply, Tie Neck and Back, Ciel Blue, XL, 12/Case
Supplier:  Med Vet International MDT012090XL
Encompass_Preferred
Blockade Surgeons' Gowns, Lightweight: Medline'S Blockade Fabric Is A Taffeta Weave Produced With Untwisted Continuous Filament Polyester. Constructed Of 99% Polyester/1% Carbon. Medline'S Blockade Fabric Provides An Exceptional Barrier That Is Highly Fluid-Resistant. The High-Density Construction Resists Fluid Penetration While Maintaining Breathability And Comfort. Under Normal Conditions, Blockade Barrier Properties Are Guaranteed Up To 75 Processings. These Comfortable Gowns Feature A 1-Ply Blockade On Full Front And Full Sleeves With Angelstat Fabric Back Panels. Bias Bound Color-Coded Neck Line Indicates The Gown'S Size. Available With Tie Neck And Back Closure. X-Large, Ceil Blue.
Catalog No. 50-209-2234
Due to product restrictions, please to purchase or view availability for this product.