Learn More
Med Vet International GOWN SURGEON STERILE XLARGE
Supplier: Med Vet International OSGXL
Full back with wrap-around style and knit cuffs. Surgical barrier fabric engineered from non-woven SMS fabric. High level of fluid repellency. Manufactured for protection, sterility and comfort during surgical procedures. Ample room in the chest and sleeves for unrestricted movement. Fully adjustable hook and loop neckline makes gowning easy.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.