Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Light chain Goat anti-Rat, Polyclonal, Secondary Antibody, MilliporeSigma™
Goat Polyclonal Antibody
Supplier: MilliporeSigma AP202
Description
Goat anti-Rat light chain Antibody is an antibody against light chain for use in ELISA and western blotting.Specifications
Goat anti-Rat light chain Antibody | |
Polyclonal | |
Purified by immunoaffinity chromatography. Liquid in 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6. | |
Rat IgG light chain. | |
1 mg | |
Rat | |
IgG |
ELISA, Western Blot | |
Unconjugated | |
Goat | |
Affinity Purified | |
Secondary | |
2-8°C Lyophilized for up to 12 months from date of receipt −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Protect from light |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction