Learn More
MilliporeSigma™ GM-CSF ELISPOT Antibody Pair, Human
Protein Interaction Antibody Pairs
Supplier: MilliporeSigma™ ELI012H
Description
Includes:
- Human GM-CSF Capture Antibody, clone BVD2-23B6. Quantity: 500 μg.
- Human GM-CSF Detection Antibody, clone BVD2-21C11. Quantity: 100 μg.
- Streptavidin Alkaline Phosphatase. Quantity: 55 μL.
Specifications
Monoclonal | |
Rat | |
Primary | |
2-8°C for up to six months | |
Purified |
1.0mg/mL (Capture antibody), 0.5mg/mL (Biotinylated detection antibody) | |
96-well Plates | |
Human | |
BVD2-23B6 (Capture), BVD2-21C11 (Detection) |