Learn More
Bioassay Systems QuantiChrom™ Glyoxalase I Assay Kit. For quantitative determination of glyoxalase activity. Note: use a UV plate (e.g. 96-well UV plate, cat# P96UV).
Supplier: Bioassay Systems DGLO100
For quantitative determination of glyoxalase enzyme activity. Key Features: Fast and sensitive. Detection limit of 4 U/L GLO-1 activity. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD240nm (Use a UV plate, e.g. 96-well UV plate, cat# P96UV). Samples: Enzyme preparations or biological samples. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 4 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.