Learn More
Medchemexpress LLC HY-22166 5mg Medchemexpress, Glycolic acid oxidase inhibitor 1 CAS:77529-42-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY221665MG
Encompass_Preferred
Medchemexpress, HY-22166 5mg Glycolic acid oxidase inhibitor 1 CAS:77529-42-1 Glycolic acid oxidase inhibitor 1 is a glycolate oxidase inhibitor, extracted from patent EP0021228A1, in Table IV. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2219
Explore available promotions