Learn More
Medchemexpress LLC HY-W015967 1g Medchemexpress, Glycolic acid CAS:79-14-1 Purity:>98%
Supplier: Medchemexpress LLC HYW0159671G
Medchemexpress, HY-W015967 1g Glycolic acid CAS:79-14-1 Glycolic acid is an inhibitor of tyrosinase , suppressing melanin formation and lead to a lightening of skin colour. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.