Learn More
Bioworld Glycine max (Soybean) Lectin (SBA) - Rhodamine (TRITC)
Supplier: Bioworld 217610252
Technical Specifications:Ligand Source:Glycine maxMolecular Weight:120 kDaCarbohydrate Specificity:GalNAc, GalNAcα1,3GalInhibiting/Eluting Sugar:N-AcetylgalactosamineCarbohydrate for Elution:0.2MN-AcetylgalactosamineBlood Group Specificity:A>O>BConjugate: RhodamineForm:LiquidIsoelectric (pI):5.8-6Storage: -20°CApplication note: Calcium and Manganese are required for binding.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.