Learn More
Bioassay Systems EnzyChrom™ Glutathione Peroxidase Assay Kit. For quantitative determination of glutathione peroxidase activity and evaluation of drug effects on GPX activity.
SDP
Supplier:  Bioassay Systems EGPX100
Encompass_Preferred
For quantitative determination of glutathione peroxidase activity and evaluation of drug effects on GPX activity. Key Features: Sensitive and accurate. Use 10 µL sample. Linear detection range 40 to 800 U/L GPX activity. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD340nm. Samples: Biological. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 40 U/L.
Catalog No. 50-489-274