Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific γ-Glutamyl Transferase Reagents for Hitachi™ Analyzers
Quantify γ-GT easily with system specific reagents
Supplier: Pointe Scientific HG959315
Includes: Reagent 1 and Reagent 2 when combined: Tris buffer (pH 8.1±0.1) 89mM; Glycylglycine 126mM; GLUPA-C 3.2mM; Sodium azide 0.095%; stabilizer
Description
- Method: Modified Szasz, Kinetic
- Format: Liquid (ready to use)
- Linearity: 800 U/L
- Expected Values: 6 to 54 U/L
Applications:
For the quantitative kinetic determination of γ-glutamyl transferase (γ-GT) activity in serum on Hitachi analyzers
Specifications
CE Marked | |
R1 and R2 contains Tris Buffer 89mM/L pH 8.1 ±0.1, Glycylglycine 126mM/L, GLUPA-C 3.2mM/L, Sodium Azide 0.095% | |
5 x 45mL R1, 2 x 45mL R2 |
gamma-Glutamyl Transferase Reagent Kit | |
Quantitative kinetic determination of gamma Glutamyl Transferase activity in Serum | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For use with in vitro diagnostics only
Spot an opportunity for improvement?
Provide Content Correction