Learn More
Bioassay Systems EnzyChrom™ Glucose Oxidase Assay Kit. For quantitative determination of glucose oxidase activity and evaluation of drug effects on its metabolism.
Supplier: Bioassay Systems EGOX100
For quantitative determination of glucose oxidase activity and evaluation of drug effects on its metabolism. Key Features: Sensitive and accurate. Use as little as 20 µL samples. Linear detection range in 96-well plate for 20 minute incubation at 25°C: 0.02 to 10 U/L glucose oxidase for colorimetric assays and 0.002 to 1.5 U/L for fluorimetric assays. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 20 min at room temperature. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD570nm, FL530/585nm. Samples: Cell/tissue lysate, cell culture media etc. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 0.02 (OD), 0.002U/L (FL).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.