Learn More
Invitrogen™ Glucose Colorimetric Detection Kit
Assay Glucose levels in Serum, Plasma, Urine, Buffers and Tissue Culture Media.
$638.00
Specifications
Content And Storage | 96-well plate(s), standard, other assay specific reagents, buffers, and lot-specific technical data sheet. Store at –20°C. |
---|---|
Detection Method | Colorimetric |
Immunoassay Kit Format | Colorimetric Detection Kit |
Product Type | Glucose Colorimetric Detection Kit |
Sample Type | Other Biological Fluids, Plasma, Serum |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EIAGLUC
|
Invitrogen™
EIAGLUC |
2 x 96 Tests |
Each of 1 for $638.00
|
|
|||||
Description
The Glucose Detection research-use-only kit is a colorimetric assay designed for the quantification and detection of glucose in serum, plasma, urine, buffers and tissue culture media.
This complete, ready-to-use kit includes clear 96-well plate(s), glucose standard, assay buffer, and other components to perform the assay. A 96-well microplate reader capable of reading optical density at 560 nm is required for use of this kit.
Performance characteristics
• Assay type: colorimetric detection kit
• Sample types: serum, plasma, urine, buffers, and tissue culture media
• Sensitivity: 0.413 mg/dL
• Standard curve range: 0.5–32 mg/dL
• Reactivity: species independent
Background
Glucose is by far the most common carbohydrate. It is a monosaccharide, an aldose, a hexose, and a reducing sugar and is also known as dextrose, because it is dextrorotatory (rotates polarized light clockwise). The structure of glucose is shown below as both the straight chain and cyclic forms. For all biological and molecular events and for multiple cellular functions, energy is essential.
Energy is available in the form of ATP (adenosine triphosphate), most of which is generated through aerobic cellular respiration of carbohydrate and glucose, the major source of biological free energy in higher organisms. Reduced energy levels threaten cellular homeostasis and integrity. Impaired energy metabolism may trigger pro-apoptotic signaling (programmed cell death), oxidative damage, excitotoxicity, and impede mitochondrial DNA repair.
Assay principle
The Glucose Colorimetric Detection Kit is designed to quantitatively measure glucose in a variety of samples. A beta-D-glucose standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Samples are mixed with the colorimetric substrate and horseradish peroxidase and the reaction initiated by addition of glucose oxidase. The reaction is incubated at room temperature for 30 minutes. The glucose oxidase reacts with glucose to produce hydrogen peroxide which, in the presence of HRP, reacts with the colorimetric substrate to convert the colorless substrate into a pink-colored product. The pink product is read at 560 nm. Increasing levels of glucose cause a linear increase in color.
Specifications
96-well plate(s), standard, other assay specific reagents, buffers, and lot-specific technical data sheet. Store at –20°C. |
|
Colorimetric Detection Kit | |
Other Biological Fluids, Plasma, Serum | |
Non-Gene | |
Approved for shipment on Wet or Dry Ice |
Colorimetric | |
Glucose Colorimetric Detection Kit | |
Microplate Reader | |
Non-Gene | |
Glucose |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.