Learn More
Revvity Health Sciences Inc Glucagon Serum Kit, 1000 Tests
Supplier: Revvity Health Sciences Inc 62SGLPEB
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The Glucagon offers superior specificity and sensitivity for a reliable quantification of glucagon in serum and plasma samples. Built on a truly homogeneous protocol, the assay does not require any washing or complex extraction steps, meaning significant time savings over ELISA methods. Simply add the detection reagents, incubate, and read!
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.