Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ GloWear™ Class 3 Zip-Up Hi-Vis Hooded Sweatshirt with Black Front
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Safety sweatshirt has two slash-style pockets for additional hand warmth and storage
- bottom front and wrist sections of the sweatshirt are black to prevent dirt from showing
- Two inch ANSI-compliant silver reflective tape on torso, shoulders and sleeves for effective visibility as night falls
- Zipper front closure, drawstring hood and elastic knit waistband and cuffs keep the heat in for all-day warmth
- Ideal for jobsites in high-traffic areas like road construction or anywhere that requires high visibility clothing or for activities such as biking, snowmobiling, snow shoeing, and ice fishing
Specifications
Size | Medium |
Material | Polyester polar fleece |
Color | Hi-vis Lime |
Certifications/Compliance | ANSI/ISEA 107-2015 Type R, Class 3 compliant |
For Use With (Application) | Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction, Railway Workers, Warehouse/Distribution |
Includes | Drawstring hood |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction