Learn More
Ergodyne™ GloWear™ 8365 Type R Class 3 High Visibility Rain Jacket
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Weatherproof 300D oxford polyester outer shell
- Breathable PU coating with polyester mesh lining
- 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
- Inset hood with drawstring and stoppers
- Two external front pockets with snap flaps
- Chest pocket
- Dual mic tabs for clipping radios
- YKK #5 zippers
Specifications
Color | Lime |
Material | 300D Oxford Polyester |
Size | 5X-Large |
No. of Pockets | 3 |