Learn More
Electron Microscopy Sciences Glazed Porcelain Plate, 12 Well
Supplier: Electron Microscopy Sciences 7157512
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Our economical glazed porcelain plate is made from high purity raw material, uniform in quality and resistant to acids and alkalis. It can withstand sudden temperature changes without cracks, explosion or deformation. Under normal conditions, the glazed plate can sustain a temperature of up to 1050 degree C.