Learn More
United Scientific Supplies Glass Vacuum Adapters
Makes connecting glassware safe and easy
$24.75
Specifications
Includes | 3/8 in. Hose Connector |
---|---|
Material | Borosilicate Glass |
Product Type | Vacuum Adapter |
Standard Taper Joint No. | 24/40 |
Description
- Includes a 3/8 in. Hose Connector
Specifications
3/8 in. Hose Connector | |
Borosilicate Glass | |
Vacuum Adapter | |
24/40 |