Learn More
Chemglass Life Sciences Extraction Thimble, Glass, Medium, X-Coarse Frit, 35mm OD X 90mm Height
Supplier: Chemglass Life Sciences CG137204
Extraction Thimble, Glass, Medium, X-Coarse Frit, 35mm OD X 90mm Height. All glass extraction thimble permits viewing during extraction process and subsequent weighing. Sizes correspond to CG-1368 or CG-1369 extractor. Extra coarse porosity. Sizes and porosities other than those listed are available upon request.