Learn More
United Scientific Supplies Glass Separatory Funnel
Glass Separatory Funnels are pear shaped for easy handling and use.
$58.25 - $102.00
Specifications
Material | Borosilicate Glass 3.3 |
---|---|
Color | Clear |
Product Type | Separatory Funnel |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
S82832
|
United Scientific Supplies
SF14950 |
50 mL |
Pack of 2 for $61.75
|
|
|||||
S82833
|
United Scientific Supplies
SF149100 |
100 mL |
Pack of 2 for $62.50
|
|
|||||
S82834
|
United Scientific Supplies
SF149125 |
125 mL |
Pack of 2 for $67.00
|
|
|||||
S82835
|
United Scientific Supplies
SF149250 |
250 mL |
Pack of 2 for $69.25
|
|
|||||
S82836
|
United Scientific Supplies
SF149500 |
500 mL |
Pack of 2 for $74.25
|
|
|||||
S82837
|
United Scientific Supplies
SF1491000 |
1000 mL |
Each for $58.25
|
|
|||||
S82838
|
United Scientific Supplies
SF1492000 |
2000 mL |
Each for $102.00
|
|
|||||
Description
- These pear-shaped borosilicate glass funnels include a PTFE stopcock and a plastic stopper.
- Feature a marking spot and large white block letters for easy identification of capacity, as per ASTM E1096.
- PTFE stopcocks are compliant with ASTM standard E675.
Specifications
Borosilicate Glass 3.3 | |
Clear | |
Separatory Funnel |