Learn More
MilliporeSigma™ Supelco™ Glass Packed GC Column (fits Agilent, Configuration A), Phase: None, Matrix 80/100 Porapak QS Support
Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness.
Supplier: MilliporeSigma™ Supelco™ 13094U
Description
Porapak QS is a cross-linked polyaromatic resin which is capable to extract organic compounds and any trace metals associated with organic molecules.Specifications
Packed GC | |
Gas Chromatography (GC) | |
Glass Column | |
2 mm |
0.25 in. | |
1.8 m | |
Porapak QS |