Learn More
Eisco™ Glass Burette with Needle Valve Stopcock, Sure Trace Certification
Class A, DIN 385 Compliant, Borosilicate Glass with PTFE Needle Valve Stopcock, with graduations.
$15.75 - $20.50
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
12-183-073
|
Eisco™
CH0238AST |
10 mL |
Each for $15.75
|
|
|||||
12-183-072
|
Eisco™
CH0238BST |
25 mL |
Each for $17.50
|
|
|||||
12-183-071
|
Eisco™
CH0238CST |
50 mL |
Each for $20.50
|
|
|||||
12-183-070
|
Eisco™
CH0238DST |
100 mL |
Each for $16.25
|
|
|||||
Description
- Accurate as per class A, DIN ISO 385 Compliant
- Includes an attached PTFE needle valve rotaflow stopcock
- Burette is made of precisely bore high quality, heavy duty Borosilicate 3.3 glass
- Features chemical resistant blue printing - All contents are included in a single package
Specifications
Borosilicate Glass (3.3) | |
Buret | |
Class A |