Learn More
World Precision Instrument Multi-Barrel Capillary Glass-2B150F-6
Supplier: World Precision Instrument 2B150F6
Borosilicate glass capillaries: Close dimensional tolerances assure microelectrode uniformity and reproducibility. Capillaries are available in 1, 2, 3, 5 and 7-barrel configurations, complete range of single barrel thin-wall sizes and a variety of special configurations. Capillaries with filaments contain a solid filament fused to the inner wall, which speeds filling of electrodes. Capillaries with or without inner filaments are available for making microelectrodes in a wide range of diameters.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.