Learn More
TARGETMOL CHEMICALS INC GJ071 OXALATE 5MG
Supplier: TARGETMOL CHEMICALS INC T31932
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GJ071 oxalate is a Nonsense suppressor. Nonsense suppressors (NonSups) induce “readthrough” i.e. the selection of near cognate tRNAs at premature termination codons and insertion of the corresponding amino acid into nascent polypeptide. purity: 100%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.