Learn More
Gilson™ PIPETMAN™ M Pipettes
Single or multichannel. Equipped with motorized piston that virtually eliminates pipetting variability among users and minimizes RSI (Repetitive Strain Injuries) among users
Supplier:  Gilson™ F81016
 View more versions of this product
Catalog No. F81016G
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Motorized piston with adjustable piston speed that is compatible with varying solution densities
  • Lockable volume to prevent inaccuracies
  • Color-coded push-button corresponds to matching tip packs
  • Service alert reminders
  • Unique tip holder design allows for a secure fit of most brands of tips
  • Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
  • Pipette can be used while charging eliminating bottlenecks and downtime
  • Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
  • Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
  • Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
Specifications
Specifications
D5000
P5000M
Pipetman™
500 to 5000 μL
1 Year
Purple
1
Pipette
Ergonomic
Product Suggestions
Product Suggestions