Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibson Bioscience Reagents: Oxidase Test
For the detection of oxidase producing organisms.
$97.00
Specifications
Formulation | N-N-N-tetramethyl-p-phenylenediamine 10g, ascorbic acid 1g, deionized water 1L |
---|---|
Storage Requirements | 2° to 25°C |
Test Time | 10 sec. |
Catalog Number | Mfr. No. | Product Type | Kit Size | For Use With (Application) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Kit Size | For Use With (Application) | Price | Quantity & Availability | |||
21-200-303
|
Gibson Laboratories LLC
60500LEX |
Oxidase Test Swab | 50 Tests | Rapid colorimetric test for presumptive identification of cytochrome oxidase activity in microorganisms |
Pack of 50 for $97.00
|
|
|||
Description
- Features a rapid colorimetric test
- Used for presumptive identification of cytochrome oxidase activity in microorganisms (Neisseria)
- Helps in identification of Pseudomonas and other cytochrome oxidase-positive gram negative microorganisms
Specifications
N-N-N-tetramethyl-p-phenylenediamine 10g, ascorbic acid 1g, deionized water 1L | |
2° to 25°C | |
10 sec. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title