Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibson Bioscience Fungal Serology: Antigens and Antisera

Supplied in 1 or 2mL vials
$476.00
Specifications
For Use With (Application) | Serodiagnostic identification procedures |
---|---|
Kit Size | 1 Vial |
Catalog Number | Mfr. No. | Product Type | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Volume (Metric) | Price | Quantity & Availability | ||||
21-200-352
![]() |
Gibson Laboratories LLC
200140LEX |
Blastomyces dermatitidis Antiserum (A) | 2mL |
Each for $476.00
|
|
21-200-355
![]() |
Gibson Laboratories LLC
200135LEX |
Histoplasma capsulatum Antiserum | 2mL |
Each for $476.00
|
|
Specifications
Serodiagnostic identification procedures | |
1 Vial |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title