Learn More
Gibco™ Embryonic Stem Cell FBS, Qualified, One Shot™ Format, US Origin
Secure sera for cell culture with least viral risk sera from BSE-free regions
$170.00 - $1012.00
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Fetal |
Country of Origin | United States |
Form | Liquid |
Format | Bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-141-079
![]() |
Gibco™
16141079 |
500 mL |
Each for $1,012.00
|
|
16-141-002
![]() |
Gibco™
16141002 |
50 mL |
Each for $170.00
|
|
Description
- Sera Category: Specialty
- Origin: United States
- Specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells
- Endotoxin level: ≤50 EU/mL (levels routinely ≤ 10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
Cell Culture, Embryonic Stem Cell Culture, Mammalian Cell Culture, Stem Cell Culture, Stem Cell Research
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
United States | |
Bottle | |
Standard (Sterile-filtered) | |
Sterile | |
Frozen |
Fetal | |
Liquid | |
Fetal Bovine Serum | |
Cattle/Bovine | |
ES Cell-Qualified |
For In Vitro Diagnostic Use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.