Learn More
Cole-Parmer Environmental Express GhostWipe SC4200 Moist Wipe, 15 x 15 cm, Individually Sealed; 200/Pk
Supplier: Cole-Parmer 3520872
The Ghostwipe completely dissolves during the digestion procedure thus allowing everything into solution. Dependent upon the material wiped, filtration may not be necessary. In addition, the Ghostwipe is more rugged than other traditional paper wipes and will not fall apart during the wiping of rough surfaces.