Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Graham-Field™ Grafco™ Disposable Penlites
Compact, lightweight and ready to use
$32.25
Specifications
Light Source | Low-drain LED |
---|---|
For Use With (Application) | Compact, lightweight and ready to use, deliver high-intensity bright light without the inconvenience of changing batteries or bulbs, for the physician, nurse, technician or paramedic |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-027480
|
Graham-Field™
4062 |
Disposable Diagnostic Penlight |
Pack of 6 for $32.25
|
|
|||||
Description
Deliver high-intensity bright light without the inconvenience of changing batteries or bulbs.
- Ideal for the physician, nurse, technician or paramedic
- Bright-white, low-drain LED
- Latex-free
- Shelf life of one year
Specifications
Low-drain LED | |
Compact, lightweight and ready to use, deliver high-intensity bright light without the inconvenience of changing batteries or bulbs, for the physician, nurse, technician or paramedic |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title