Learn More
Getinge USA ChemiSix Emulator Test Pack 40 tests/pk
Supplier: Getinge USA 61301606235
Contains a chemical integrator test card that meets the requirements for ANSI/AAMI/ISO 11140-1:2005 Class 6 emulators. The instant readout emulator card is contained within a pack of porous and nonporous materials that provide resistance to the penetration of steam during the sterilization cycle. Features: Reliable, instant visual results, Pre-assembled and ready-to-use, Performance proven with timing beyond biological kill, Lead free, Emulator card can be used as permanent record
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.