Learn More
Germaine™ Laboratories Brucella Melitensis Antigen
Febrile Antigens intended for detection of bacterial agglutinins associated with Brucella Melitensis infection
Supplier: Germaine™ Laboratories 1002
Description
- 5mL vial
Specifications
Brucella melitensis Antiodies | |
Brucella melitensis Antigen | |
5 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For professional use only