Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Germaine™ Laboratories Febrile Antigens For Proteus OX
Febrile Antigens intended for detection of bacterial agglutinins associated with Proteus OX infection
$56.25
Catalog Number | Mfr. No. | Detectable Analytes | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detectable Analytes | Type | Price | Quantity & Availability | ||||
16-414-105
|
Germaine™ Laboratories
1200 |
Proteus OXK Antibodies | Proteus OXK Antigen |
Each for $56.25
|
|
||||
16-414-106
|
Germaine™ Laboratories
1202 |
Proteus OX19 Antibodies | Proteus OX19 Antigen |
Each for $56.25
|
|
||||
16-414-104
|
Germaine™ Laboratories
1201 |
Bacterial Agglutinins associated with a bacterial infection | Proteus OX2 Antigen |
Each for $56.25
|
|
||||
Description
- 5mL vial
Specifications
5 mL |
Professional use only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title