Learn More
Geotech Environmental Equiptment Inc Geotech Sand Shaker
Supplier: Geotech Environmental Equiptment Inc 81450001
The Geotech Sand Shaker is a mechanical sieve kit designed to provide reliable grain size analysis. It features 20 stainless steel screens ranging in size from US sieve No. 4 to 270. The sample is placed into the top of five clear acrylic cylinders and shaken to distribute particles. Volumetric percentages are indicated on the Sand Shaker frame.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.