Learn More
CROSSTEX INTERNATIONAL 1 mL Spore Ampules, 10^6, 50/pk
Supplier: CROSSTEX INTERNATIONAL NMSA15006
Crosstex 1 mL Spore Ampoules are Self-Contained Biological Indicators for monitoring steam processes consisting of a hermetically sealed glass ampoule which contains modified Trypticase Soy Broth (TSB) with a pH indicator and Geobacillus stearothermophilus spores with a population level of 10^4, 10^5 or 10^6.