Learn More
Metrohm USA GENRATR W/DIA F/756 6.0344.100
Supplier: Metrohm USA 60344100
Generator electrode with diaphragm. Generator electrode for Karl Fischer titrations, with diaphragm, standard ground-joint 29/22. Electrode plug-in head: Metrohm plug-in head G. Indicator electrode type:Cathode = Pt grid. Indicator electrode type (reference):Anode = Pt grid. Min. immersion depth (mm):15. SGJ sleeve:Standard ground-joint 29/22. Shaft diameter bottom (mm):24. Shaft diameter top (mm):24. Shaft material:Glass. Temperature range, long-term ( Deg.C):-20 ... 70. Temperature range, short-term ( Deg.C):-20 ... 70.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.