Learn More
Genetic Traits Test Strips
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
A quick, easy, and completely safe means to analyze commonly tested genetic traits.
Supplier: Neo/Sci Corporation 992405
Description
Simple taste tests allow students to determine their possible genotypes for the trait and the frequency of tasters and non-tasters in a population, as well as construct a family pedigree showing the heredity of the trait.
Four types available:
- PTC taste test strips
- Sodium benzoate taste test strips
- Thiourea taste test strips
- Control taste test strips
Specifications
Control Taste Strips | |
To determine their possible genotypes for the trait and the frequency of tasters and non-tasters in a population, as well as construct a family pedigree showing the heredity of the trait |