Learn More
Applied Biosystems™ GeneScan™ 500 TAMRA™ dye Size Standard
GeneScan™ 500 TAMRA™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence- based DNA electrophoresis systems.
Supplier: Applied Biosystems™ 401733
Includes: This kit contains 16 TAMRA dye labeled single stranded DNA oligos for size standards.
Description
- The use of an internal lane size standard during electrophoresis enables automated data analysis and precise DNA fragment size comparisons between electrophoresis runs
Order Info
Each
Specifications
GeneScan™, TAMRA™ | |
310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer | |
Wet Ice |
This kit contains 16 TAMRA dye labeled single stranded DNA oligos for size standards. Store kit at 2-8°C. (Do not freeze) | |
TAMRA™ |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.