Learn More
Diversified Spaces™ General Storage Cabinet
Used for general storage of items.
Supplier: Diversified Spaces™ P7152BK36SC
Description
- Solid hardwood, solid maple and maple veneers construction
- Earth-friendly UV finish
- Hinged maple doors
- 3-point locking system
- Shelf Height: 1 in. (For 36 in. wide cabinet), 0.75 in. ( For 24, 30 in. wide cabinets)
Certifications
MAS Certified Green
Compliance
SEFA
Warranty and Services
Limited Lifetime Warranty
Specifications
76.2 cm | |
55.88 cm | |
84 in. | |
22 in. |
Storage Cabinet | |
213.36 cm | |
30 in. | |
Storage of general supplies |