Learn More
Perkin Elmer US LLC General Purpose FT-IR Sampling Kit
Supplier:  Perkin Elmer US LLC L1365312
Encompass_Preferred
This kit contains all of the necessary equipment for handling liquids, mulls and KBr pellets in a rugged storage case. Its purpose is to supply the newcomer to FT-IR spectroscopy with the more frequently required items in FT-IR analysis.
Catalog No. 50-211-7210