Learn More
Thermo Scientific™ GeneJET™ Plasmid Miniprep Kit Promotion
Perform isolation of high quality plasmid DNA from recombinant &italic_begin;E.coli&italic_end; cultures with this simple, rapid and cost-effective system.
Supplier: Thermo Scientific™ K0503
Description
GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column. The kit recovers up to 20 μg of high copy plasmid DNA per isolation procedure.
Highlights
- Efficient -high yields of up to 20 µg of high quality plasmid DNA
- Fast -procedure takes less than 14 minutes
- Convenient -no phenol/chloroform extraction or alcohol precipitation required
- Pure -purified DNA is immediately ready to use
Specifications
Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:—FastDigest or conventional restriction digestion,—automated fluorescent and radioactive sequencing | |
GeneJET Plasmid Miniprep Kit |