Learn More
Nasco Gen II Ultrasound Central Line Training Model
Gen II Ultrasound Central Line Training Model is a training tool for ultrasound applications for the torso venous system.
Supplier: Nasco BPH660HP
Description
- Visualize the pulsatility of arteries via ultrasound for positive identification of the targeted vessel
- Realistic anatomy of the upper thorax and neck
- Self-healing tissue offers users tremendous durability
- Venous anatomy includes internal jugular vein (IF), brachiocephalic vein, subclavian vein, and axillary vein
- Simulated superior vena cava, right atrium, and right ventricle offer users the ability to fully thread guide wires, dilators, and catheters without resistance
- Arterial pulsation can be simulated via the manual hand bulb
- Optional automated pump allows users to differentiate between arteries and veins
Specifications
Medical Training Model | |
Ultrasound Training | |
Tan |