Learn More
Gemini™ Chemical Identification Analyzers
Identify unknown chemicals and explosives in the field quickly, safely, and confidently with the power of FTIR and Raman spectroscopy in a single handheld instrument.
$128600.00 - $164500.00
Specifications
Certifications/Compliance | Certified to MIL-STD-810G and IP67; full test report available on request |
---|---|
Data Export Formats | SPC file (for use in standard spectroscopic software), text file, or JPEG report |
Description | Gemini Analyzer |
IP Rating | IP67 |
Languages | English |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
18-001-145
|
Thermo Scientific
8000501101 |
1 year of support: reachback support, software updates and library updates |
Each for $128,600.00
|
|
|||||
18-001-146
|
Thermo Scientific
8000501201 |
2 years of support: reachback support, software updates and library updates |
Each for $137,600.00
|
|
|||||
18-001-147
|
Thermo Scientific
8000501301 |
3 years of support: reachback support, software updates and library updates |
Each for $148,900.00
|
|
|||||
18-001-148
|
Thermo Scientific
8000501401 |
4 years of support: reachback support, software updates and library updates |
Each for $156,600.00
|
|
|||||
18-001-149
|
Thermo Scientific
8000501501 |
5 years of support: reachback support, software updates and library updates |
Each for $164,500.00
|
|
|||||
Description
Identify a broad range of unknown chemicals and explosives in the field using FTIR and Raman spectroscopy in a single instrument, including low concentration analysis. The rugged, compact Thermo Scientific™ Gemini™ analyzer meets the demanding requirements of elite military and public safety forces, helping operators execute their mission quickly, safely, and accurately.
The Gemini Analyzer is the first and only integrated Raman and FTIR handheld instrument in the world, providing complementary and confirmatory testing in a single, field-portable device.
Better, faster decision making
- Lightweight and compact, ideal for chemical battalions, explosive ordnance disposal (EOD) technicians and other first responders burdened with heavy equipment.
- Virtual mixture analysis and interactive problem solving on scene with HazMasterG3.
- Includes 24/7 reachback support with access to Ph.D. spectroscopists for spectral analysis and operational inquiries.
Increased confidence in results
- Leverages both Raman and FTIR technology to address a broader range of samples than either technique alone.
- Identifies unknown solids and liquids, from explosives and chemical warfare agents to industrial chemicals and precursors using a comprehensive onboard library.
- Builds on the field-proven performance of Thermo Scientific™ FirstDefender™ and Thermo Scientific™ TruDefender™ Analyzers.
Safer operation
- Includes a motorized anvil which adjusts sample pressure based on user settings.
- Features Raman and FTIR Scan Delay to allow users to leave the hazard zone before initiating a scan.
Easier to use, enhanced efficiency
- Guides the operator through technology selection and sampling with Scan Assist.
- Customizable Profiles allow the operator to set scan parameters based on the sample, scenario and their situational awareness
- Minimizes user error and eliminates subjective user interpretation.
- Requires minimal training for proficiency.
Specifications
Certified to MIL-STD-810G and IP67; full test report available on request | |
Gemini Analyzer | |
English | |
FTIR: 4,000 cm-1 to 650 cm-1 Raman: 250 cm-1 to 2875 cm-1 |
|
4.2 lb. |
SPC file (for use in standard spectroscopic software), text file, or JPEG report | |
IP67 | |
Internal 3.7 V lithium polymer (QTY 1) battery pack Internal CR123A (QTY 3) batteries DC wall adapter, 12 V DC, 1.25 A |
|
-20°C to +50°C | |
1.9 kg |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.