Learn More
Medchemexpress LLC HY-17026 1g , Gemcitabine NSC 613327;LY188011 CAS:95058-81-4 Purity:98%
Supplier: Medchemexpress LLC HY170261G
Medchemexpress, HY-17026 1g Gemcitabine NSC 613327;LY188011 CAS:95058-81-4 Formula:C9H11F2N3O4 DNA synthesis Purity:98% Gemcitabine is a DNA synthesis inhibitor with IC 50 of 37.6, 42.9, 92.7, 89.3 and 131.4 nM in BxPC-3, Mia Paca-2, PANC-1, PL-45 and AsPC-1 cells, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.