Learn More
Medchemexpress LLC HY-17026 500mg Medchemexpress, Gemcitabine CAS:95058-81-4 Purity:>98%
Supplier: Medchemexpress LLC HY17026500MG
Medchemexpress, HY-17026 500mg Gemcitabine CAS:95058-81-4 Gemcitabine (LY 188011) is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. Gemcitabine inhibits DNA synthesis and repair, resulting in autophagyand apoptosis[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.