Learn More
Lonza™ GelBond™ Film for Agarose Gels
Transparent, flexible polyester film supports gels through electrophoresis, immunodiffusion, fixing, staining, destaining, and drying procedures. GelBond™ Film for Agarose Gels are reliable, durable and convenient.
$756.49 - $2067.47
Specifications
Product Type | Agarose Gel Film |
---|---|
For Use With (Application) | Agarose Gel electrophoresis, immunodiffusion, fixing, staining, destaining and drying procedures |
Material | Polyester |
Thickness (English) | 0.007 in. |
Thickness (Metric) | 0.02 cm |
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
BMA53734
|
Lonza™
53734 |
10 cm | 8.5 cm |
Each for $756.49
|
|
||||
BMA53780
|
Lonza™
53780 |
16.5 m | 20.3 cm |
Each for $2,067.47
|
|
||||
BMA53750
|
Lonza™
53750 |
16.5 m | 12.7 cm |
Each for $1,382.69
|
|
||||
BMA53749
|
Lonza™
53749 |
18 cm | 16 cm |
Each for $1,671.55
|
|
||||
BMA53740
|
Lonza™
53740 |
16.5 m | 10.2 cm |
Each for $1,119.08
|
|
||||
BMA53748
|
Lonza™
53748 |
20.5 cm | 11 cm |
Each for $1,740.23
|
|
||||
BMA53745
|
Lonza™
53745 |
12.5 cm | 11 cm |
Each for $1,113.02
|
|
||||
BMA53761
|
Lonza™
53761 |
25.8 cm | 12.4 cm |
Each for $1,920.01
|
|
||||
BMA53759
|
Lonza™
53759 |
24.5 cm | 12.5 cm |
Each for $1,857.39
|
|
||||
BMA53746
|
Lonza™
53746 |
15 cm | 10 cm |
Each for $1,199.88
|
|
||||
Description
- Permanently attaches to gels during casting
- Makes gels easier to handle
- Helps gels retain their original dimensions during staining and drying
- Available as precut sheets or rolls
Note:polyester films will not transmit light of less than 310nm and will fluoresce at higher wavelengths
Specifications
Agarose Gel Film | |
Polyester | |
0.02 cm |
Agarose Gel electrophoresis, immunodiffusion, fixing, staining, destaining and drying procedures | |
0.007 in. |